Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 1E7.2]
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 1E7.2]
Catalog # 89360-452
Supplier:  Genetex
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 1E7.2]
Catalog # 89360-452
Supplier:  Genetex
Supplier Number:  GTX18371

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Zeta-chain (TCR) associated protein kinase 70kDa
  • Antigen Symbol:
    ZAP70
  • Clonality:
    Monoclonal
  • Clone:
    1E7.2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    7535
  • Antigen Synonyms:
    ZAP-70|syk-related tyrosine kinase|FLJ17670|SRK|tyrosine-protein kinase ZAP-70|TZK|70 kDa zeta-associated protein|zeta-chain associated protein kinase|70kD|STD|FLJ17679
  • Amino Acid Number:
    282 to 307
  • Storage Buffer:
    Preservative: 0.09% Sodium Azide. Constituents: PBS. pH 7.2
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    GST-fusion protein: RRIDTLNSD GYTPEPARIT SPDKPRP, corresponding to amino acids 282-307 of Human ZAP70.
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Cat. No.:
    89360-452
  • Supplier no.:
    GTX18371

Specifications

About this item

ZAP70, a 70 kDa member of the Syk tyrosine kinase family, plays a central role in lymphocyte activation and development, and is implicated in several immune disorders. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling. Phosphorylation of tyrosines 315 and 319 plays an important role in mediating the positive and negative selection of T cells in thymus.Mutations in ZAP70 gene results in a form of Severe Combined Immunodeficiency Syndrome (SCID) in humans. ZAP70 expression also defines a subset of Chronic Lymphocytic Leukemia (CLL) in patients with unmutated Ig gene and poor clinical course. Recent studies suggest that protein levels of ZAP70 are elevated in B cells of CLL patients with non mutant heavy chain variable region (IgVH) but not those with the mutant regions. Recent evidence suggests that ZAP70 could be an excellent prognostic biomarker with high levels of the proteins indicating a poor prognosis.

For ELISA: Use at an assay dependent dilution. For FACS: Use at an assay dependent dilution. IF: Use 0.5?g for 10^6 cells in 100ul volume. IP: Use at an assay dependent dilution. WB: Use at a concentration of 0.5-1 ?g/ml. Detects a band of approximately 70 kDa (predicted molecular weight: 70 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: 1E7.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human