Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin E
- Antigen Symbol:IgE
- Clonality:Monoclonal
- Clone:MARE-1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Size:125 μg
- Gene ID:299351
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing 0.09% sodium azide and 50% Glycerol
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Rat IgE myeloma proteins (IR162, IR1016, IR2 and IR410).
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Immunogen affinity purified
- Cat. No.:89358-678
- Supplier no.:GTX11667
Specifications
About this item
IgE is the class of antibodies produced in the lungs, skin, and mucous membranes. It may protect against parasite invasion. But it is a major factor in allergic reactions. The antigen-specific IgE interacts with mast cells and eosinophils, triggers the release of histamine, leukotrienes and other substances that lead to the itching, sneezing and congestion of allergies - and the life threatening respiratory distress of asthma and anaphylactic shock.
Recommended Starting Dilutions:FACS: Use at a concentration of 5 µg/mL. Use 50 µL of the suggested working dilution to label 2x10^7 cells in 100 uL. Not tested in other applicationsOptimal dilutions/concentrations should be determined by the researcher.
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat