Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TUBB Mouse Monoclonal Antibody [clone: SAP.4G5]
Anti-TUBB Mouse Monoclonal Antibody [clone: SAP.4G5]
Catalog # 89357-336
Supplier:  Genetex
Anti-TUBB Mouse Monoclonal Antibody [clone: SAP.4G5]
Catalog # 89357-336
Supplier:  Genetex
Supplier Number:  GTX11312

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tubulin beta
  • Antigen Symbol:
    TUBB
  • Clonality:
    Monoclonal
  • Clone:
    SAP.4G5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Bovine
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Format:
    Ascites
  • Gene ID:
    203068
  • Antigen Synonyms:
    tubulin beta polypeptide|tubulin beta-5 chain|M40|beta Ib tubulin|tubulin|beta 5-tubulin|tubulin beta chain|tubulin beta-1 chain|CDCBM6|beta polypeptide|beta-4 tubulin|OK/SW-cl.56|TUBB1|TUBB5
  • Storage Buffer:
    Preservative: 15mM Sodium Azide; Constituents: Raw Ascites
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide corresponding to the C-terminal sequence of beta-tubulin isotype I, coupled to BSA.
  • Tested Applications:
    ELISA, ICC/IF, IHC, WB
  • Purification:
    Ascites
  • Cat. No.:
    89357-336
  • Supplier no.:
    GTX11312

Specifications

About this item

Mouse monoclonal antibody [SAP.4G5] to beta Tubulin I

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution (methanolacetone and 3% paraformaldehyde-0.5% Triton-x-100 fixations). IHC: Use at an assay dependent dilution (10% neutral buffered formalin-alcoholic formalin-xylene, paraffin-embedded). WB: Use at a dilution of 1/20000, this dilution is determined using cultured chicken fibroblasts. Predicted molecular weight: 55kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TUBB
Clonality: Monoclonal
Clone: SAP.4G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Dog, Rat, Hamster, Xenopus laevis, Bovine