Anti-HSPA4 Rabbit Polyclonal Antibody
Catalog # 89357-056
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Heat Shock 70kDa Protein 4
- Antigen Symbol:HSPA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:3308
- Antigen Synonyms:HS24/P52|HSPH2|APG-2|HEL-S-5a|hsp70|hsp70RY|RY
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:94 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human HSP70 4. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89357-056
- Supplier no.:GTX112329
Specifications
About this item
Rabbit Polyclonal antibody to HSP70 4 (heat shock 70kDa protein 4)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: HSPA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human