Anti-COMMD7 Rabbit Polyclonal Antibody
Catalog # 89356-966
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:COMM Domain Containing 7
- Antigen Symbol:COMMD7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:149951
- Antigen Synonyms:COMMD7|dJ1085F17.3|C20orf92|COMM domain-containing protein 7
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:23 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human COMMD7. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89356-966
- Supplier no.:GTX112076
Specifications
About this item
Rabbit Polyclonal antibody to COMMD7 (COMM domain containing 7)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: COMMD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: