Anti-PEX19 Rabbit Polyclonal Antibody
Catalog # 89356-438
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Peroxisomal Biogenesis Factor 19
- Antigen Symbol:PEX19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:5824
- Antigen Synonyms:D1S2223E|peroxin-19|peroxisomal farnesylated protein|PXMP1|housekeeping gene|33 kDa housekeeping protein|33kD|PMPI|HK33|PXF|PMP1
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:33 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human PEX19. The exact sequence is proprietary.
- Tested Applications:ICC/IF, IHC-P, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89356-438
- Supplier no.:GTX110721
Specifications
About this item
Rabbit Polyclonal antibody to PEX19 (peroxisomal biogenesis factor 19)
Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PEX19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human