Anti-ACY1 Rabbit Polyclonal Antibody
Catalog # 89356-284
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Aminoacylase 1
- Antigen Symbol:ACY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:95
- Antigen Synonyms:EC 3.5.1.14|ACY1_HUMAN.|ACY1D|ACYLASE|N acyl L amino acid amidohydrolase
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:46 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Aminoacylase 1. The exact sequence is proprietary.
- Tested Applications:ICC/IF, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89356-284
- Supplier no.:GTX110348
Specifications
About this item
Rabbit polyclonal antibody to Aminoacylase 1
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: ACY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human