Specifications
- Antibody Type:Primary
- Antigen Name:Neuropeptide Y
- Antigen Symbol:NPY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Cat
- Size:50 μl
- Gene ID:24604
- Antigen Synonyms:NPY|PYY4|OTTHUMP00000201947
- Storage Buffer:Whole antiserum containing 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic Neuropeptide Y (porcine) conjugated to KLH.
- Tested Applications:Dot Blot, Radio Immunoassay
- Purification:Whole antiserum
- Cat. No.:89356-058
- Supplier no.:GTX10980
Specifications
About this item
Rabbit polyclonal to Neuropeptide Y
Dot: Use at a dilution of 1/10000. This concentration was determined using Neuropeptide Y-BSA (0.03-0.25 ng/dot). IHC-Fr: Use at a dilution of 1/8000. This concentration was determined using frozen sections of rat brain perfusion-fixed with 4% paraformaldehyde. Absorption studies show that specific staining of tissue may be inhibited following preincubation of the antiserum with 10 ?M neuropeptide Y (porcine). RIA: Use at a dilution of 1/10000. This concentration was determined using 20 pg/tube of ^125I-labeled neuropeptide Y in a second antibody and polyethylene glycol RIA. The sensitivity of the RIA assay is defined as the 90% intercept of a B/B0 standard curve. In this system the sensitivity has been found to be 20 pg/tube. The affinity constant (Ka) is determined by a Scatchard plot using this RIA system. Ka = Minimum 5.0 x 10^10 L/moleNot tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Npy
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey, Pig, Rat, Cat, Rabbit, Bovine