Anti-GPR52 Rabbit Polyclonal Antibody
Catalog # 89355-300
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor 52
- Antigen Symbol:GPR52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 μl
- Gene ID:9293
- Antigen Synonyms:GPR52_HUMAN|axor10|G protein coupled receptor 52|GPCR 52.|Probable G-protein coupled receptor 52|GPR52|Probable G protein coupled receptor 52
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:41 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1.02mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GPR52. The exact sequence is proprietary.
- Tested Applications:ELISA-Ag
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89355-300
- Supplier no.:GTX108123
Specifications
About this item
Rabbit polyclonal antibody to GPR52 (G protein-coupled receptor 52)
Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: GPR52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse