Anti-HTR1A Rabbit Polyclonal Antibody
Catalog # 89350-490
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:5-Hydroxytryptamine (Serotonin) Receptor 1A
- Antigen Symbol:HTR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:3350
- Antigen Synonyms:ADRB2RL1|5-HT1A|ADRBRL1|5-HT-1A|PFMCD|5HT1a|G-21
- Storage Buffer:1XPBS, 40% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:46 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human 5HT1A Receptor. The exact sequence is proprietary.
- Tested Applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89350-490
- Supplier no.:GTX104703
Specifications
About this item
Rabbit Polyclonal antibody to 5-HT1A receptor
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: HTR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human