Anti-ORC2 Rabbit Polyclonal Antibody
Catalog # 89349-936
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Origin Recognition Complex, Subunit 2
- Antigen Symbol:ORC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4999
- Antigen Synonyms:Origin recognition complex subunit 2|Origin of replication 2|ORC2L
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:66 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.45mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human ORC2. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89349-936
- Supplier no.:GTX103280
Specifications
About this item
Rabbit polyclonal antibody to ORC2 (origin recognition complex, subunit 2-like (yeast))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: ORC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human