Anti-HSP90B1 Rabbit Polyclonal Antibody
Catalog # 89349-912
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Heat shock protein 90kDa beta member 1
- Antigen Symbol:HSP90B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7184
- Antigen Synonyms:tumor rejection antigen 1|glucose regulated protein|ECGP|endoplasmin|GP96|GRP-94|94 kDa|heat shock protein 90 kDa beta member 1|Tumor rejection antigen-1|94 kDa glucose-regulated protein|GRP94|TRA1|endothelial cell (HBMEC) glycoprotein|gp96 homolog
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:92 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the C-terminus region of human GRP94. The exact sequence is proprietary.
- Tested Applications:ICC/IF, IHC-P, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89349-912
- Supplier no.:GTX103232
Specifications
About this item
Rabbit Polyclonal antibody to GRP94 (heat shock protein 90kDa beta (Grp94), member 1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: HSP90B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human