Anti-Y Mouse Monoclonal Antibody [clone: PY20]
89349-904
: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Tyrosine
- Antigen Symbol:Y
- Clonality:Monoclonal
- Clone:PY20
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Multiple
- Western Blot:Yes
- Antigen Synonyms:p-Tyr|Phosphotyrosine|tyr|Phospho tyr|p-tyrosine
- Modification:phosphotyrosine
- Storage Buffer:20 mM phosphate buffered saline, pH 7.5, containing 50% glycerol and 3 mM sodium azide
- Storage Temperature:Store at -0°C to –20 °C. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Recommend determining optimal working dilutions by titration test.
- Immunogen:Phosphotyrosine conjugated to KLH.
- Tested Applications:Affinity Purification
- Purification:Protein A purified
- Cat. No.:89349-904
- Supplier no.:GTX10321
This is a standard clone used to detect phosphotyrosine.
Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone: PY20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Many Species