Anti-DMC1 Rabbit Polyclonal Antibody
89349-844
: Genetex
- Antibody Type:Primary
- Antigen Name:DNA Meiotic Recombinase 1
- Antigen Symbol:DMC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:11144
- Antigen Synonyms:disrupted meiotic cDNA 1 homolog|HsLim15|DMC1H|dJ199H16.1|Meiotic recombination protein DMC1/LIM15 homolog|DMC1 homologue|DMC 1
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:38 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.9mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human DMC1. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89349-844
- Supplier no.:GTX103062
Rabbit polyclonal antibody to DMC1 (DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination (yeast))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: DMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human