Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ASIC3 Guinea Pig Polyclonal Antibody
Anti-ASIC3 Guinea Pig Polyclonal Antibody
Catalog # 89350-034
Supplier:  Genetex
Anti-ASIC3 Guinea Pig Polyclonal Antibody
Catalog # 89350-034
Supplier:  Genetex
Supplier Number:  GTX10354
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Acid Sensing Ion Channel Subunit 3
  • Antigen Symbol:
    ASIC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Guinea pig
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Size:
    50 μl
  • Gene ID:
    286920
  • Antigen Synonyms:
    Testis sodium channel 1|Amiloride sensitive cation channel 3|hTNaC1|hTNaC 1|ACCN 3|ASICs3|Neuronal amiloride-sensitive cation channel 3|DRASIC|ACCN3|ASIC3|ASIC 3|hASIC 3|Amiloride-sensitive cation channel 3|Proton gated cation channel subunit|Modulatory subunit of ASIC 2a|Acid sensing ion channel 3|Neuronal amiloride sensitive cation channel 3|SLNAC 1|Modulatory subunit of ASIC2a|Amiloride sensitive cation channel 3 testis|Acid-sensing ion channel 3|ASIC3_HUMAN|TNaC 1|hASIC3|SLNAC1|TNaC1.
  • Storage Buffer:
    Preservative: 0.05% Sodium Azide; Constituents: 1% BSA, PBS. pH 7.4
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide: TASLDPDDFDPEPSDPLGSP, corresponding to amino acids 285-304 of extracellular domain of Rat ASIC3.
  • Tested Applications:
    ICC/IF, IHC-Fr
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    89350-034
  • Supplier no.:
    GTX10354

Specifications

About this item

Guinea pig polyclonal to ASIC3

ICC: Use at a dilution of 1/100. IHC-Fr: Use at a dilution of 1/100. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ASIC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype: IgG
Reactivity: Rat