Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Sorbitol Dehydrogenase (from E. coli)
Human Recombinant Sorbitol Dehydrogenase (from E. coli)
Catalog # 102174-020
Supplier:  Novus Biologicals
CAS Number:  
undefined
Human Recombinant Sorbitol Dehydrogenase (from E. coli)
Catalog # 102174-020
Supplier:  Novus Biologicals
Supplier Number:  NBP1-44460
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 μg
  • Storage Conditions:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Enzyme Name:
    Sorbitol Dehydrogenase
  • Enzyme Synonyms:
    EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1
  • Purity:
    >90%, by SDS-PAGE
  • Concentration:
    0.5 mg/ml
  • Tested Applications:
    ELISA
  • Cat. No.:
    102174-020
  • Supplier no.:
    NBP1-44460

Specifications

About this item

The Recombinant Human Sorbitol Dehydrogenase Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Sorbitol Dehydrogenase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

  • Entrez Gene ID: 6652

Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.