Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 3
- Antigen Symbol:VAP-1/AOC3
- Clonality:Monoclonal
- Clone:32325
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Size:0.025 mg
- Gene ID:8639
- Antigen Synonyms:Vascular adhesion protein 1|EC 1.4.3|Semicarbazide-sensitive amine oxidase|VAP-1membrane primary amine oxidase|copper containing 3 (vascular adhesion protein 1)|HPAOSSAO|VAP1EC 1.4.3.21|Copper amine oxidase|amine oxidase
- Storage Buffer:0.2 um filtered solution in PBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.1 mg/ml
- Immunogen:Vessels from mouse lymph nodes
- Tested Applications:Functional
- Purification:Protein A or G purified
- Cat. No.:102191-400
- Supplier no.:NBP1-58374
Specifications
About this item
The VAP-1 / AOC3 Antibody (7-88) from Novus Biologicals is a rat monoclonal antibody to VAP-1 / AOC3. This antibody reacts with mouse. The VAP-1 / AOC3 Antibody (7-88) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional.
Type: Primary
Antigen: VAP-1/AOC3
Clonality: Monoclonal
Clone: 32325
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse