Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Pan Endothelial Cell Marker
- Antigen Symbol:Pan Endothelial Cell Marker
- Clonality:Monoclonal
- Clone:RM0061-4J13
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2
- Reactivity:Mouse
- Size:100 μg
- Form:Lyophilized
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Cell lysates of mouse endothelial cells
- Tested Applications:, CyTOF-ready
- Purification:Protein G purified
- Cat. No.:102251-538
- Supplier no.:NBP2-12438
The Pan Endothelial Cell Marker Antibody (RM0061-4J13) from Novus Biologicals is a rat monoclonal antibody to Pan Endothelial Cell Marker. This antibody reacts with mouse. The Pan Endothelial Cell Marker Antibody (RM0061-4J13) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
Type: Primary
Antigen: Pan Endothelial Cell Marker
Clonality: Monoclonal
Clone: RM0061-4J13
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse