Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin specific peptidase 26
- Antigen Symbol:USP26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:83844
- Antigen Synonyms:EC 3.4.19.12|MGC120067|Deubiquitinating enzyme 26|ubiquitin-specific processing protease 26|Ubiquitin-specific-processing protease 26|MGC120066|MGC120068|ubiquitin carboxyl-terminal hydrolase 26|ubiquitin specific peptidase 26|ubiquitin specific protease 26|ubiquitin thioesterase 26|Ubiquitin thiolesterase 26
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to USP26 (ubiquitin specific peptidase 26) The peptide sequence was selected from the middle region of USP26 (NP_114113). Peptide sequence DILDKIKSKAKETKRNDDKGDHTYRLISVVSHLGKTLKSGHYICDAYDFE.
- Purification:Immunogen affinity purified
- Cat. No.:102198-576
- Supplier no.:NBP1-69152
Specifications
About this item
The USP26 Antibody from Novus Biologicals is a rabbit polyclonal antibody to USP26. This antibody reacts with human. The USP26 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: USP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human