Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 102130-954
  • 102130-966
  • Antigen Symbol:
  • UCH-L1/PGP9.5
  • UCH-L1/PGP9.5
  • Antigen Name:
  • Ubiquitin Carboxyl-Terminal Esterase L1
  • Ubiquitin Carboxyl-Terminal Esterase L1
  • Antigen Synonyms:
  • Neuron cytoplasmic protein 9.5|EC 3.4.19.12|PGP9.5|UCHL1|PGP9.5Uch-L1|ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|ubiquitin carboxyl-terminal hydrolase isozyme L1|ubiquitin C-terminal hydrolase|PGP95|EC 6.-|PARK5
  • Neuron cytoplasmic protein 9.5|EC 3.4.19.12|PGP9.5|UCHL1|PGP9.5Uch-L1|ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|ubiquitin carboxyl-terminal hydrolase isozyme L1|ubiquitin C-terminal hydrolase|PGP95|EC 6.-|PARK5
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 7345
  • 7345
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • A synthetic peptide made to an internal portion of the human UCHL1 protein (between residues 100-200). [UniProt# P09936]
  • A synthetic peptide made to an internal portion of the human UCHL1 protein (between residues 100-200). [UniProt# P09936]
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage Buffer:
  • Tris-Glycine and 0.15M NaCl
  • Tris-Glycine and 0.15M NaCl
  • Storage Temperature:
  • Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

Specifications

Related Information