Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody (Biotin)
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody (Biotin)
Catalog # 102130-970
Supplier:  Novus Biologicals
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody (Biotin)
Catalog # 102130-970
Supplier:  Novus Biologicals
Supplier Number:  NB300-676B

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Ubiquitin Carboxyl-Terminal Esterase L1
  • Antigen Symbol:
    UCH-L1/PGP9.5
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    7345
  • Antigen Synonyms:
    Neuron cytoplasmic protein 9.5|EC 3.4.19.12|PGP9.5|UCHL1|PGP9.5Uch-L1|ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|ubiquitin carboxyl-terminal hydrolase isozyme L1|ubiquitin C-terminal hydrolase|PGP95|EC 6.-|PARK5
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    A synthetic peptide made to a C-terminal portion of the human UCHL1 protein (between residues 200-220). [UniProt# P09936]
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102130-970
  • Supplier no.:
    NB300-676B

Specifications

About this item

The UCH-L1 / PGP9.5 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to UCH-L1 / PGP9.5. This antibody reacts with human, mouse. The UCH-L1 / PGP9.5 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: UCH-L1/PGP9.5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse