Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Twik-Related K+ Channel 1
- Antigen Symbol:TREK 1
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Bovine,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3776
- Antigen Synonyms:tandem-pore-domain potassium channel TREK-1|subfamily K|TWIK-related potassium channel 1|MGC126744|two-pore potassium channel 1|TREK-1 K(+) channel subunit|member 2|potassium channel|MGC126742
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2]
- Purification:Immunogen affinity purified
- Cat. No.:102122-296
- Supplier no.:NB110-41535R
The TREK 1 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to TREK 1. This antibody reacts with human, mouse, bovine. The TREK 1 Antibody [DyLight 550] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: TREK 1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine