Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Teashirt homolog 1
- Antigen Symbol:TSHZ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10194
- Antigen Synonyms:SDCCAG33|teashirt homolog 1|Serologically defined colon cancer antigen 33teashirt family zinc finger 1|Antigen NY-CO-33|TSH1teashirt zinc finger 1|NY-CO-33|teashirt zinc finger homeobox 1
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SDCCAG33. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102165-748
- Supplier no.:NBP1-32300
Specifications
About this item
The Teashirt homolog 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Teashirt homolog 1. This antibody reacts with human. The Teashirt homolog 1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Teashirt homolog 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human