Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TBC1D26 Rabbit Polyclonal Antibody
Anti-TBC1D26 Rabbit Polyclonal Antibody
Catalog # 102190-198
Supplier:  Novus Biologicals
Anti-TBC1D26 Rabbit Polyclonal Antibody
Catalog # 102190-198
Supplier:  Novus Biologicals
Supplier Number:  NBP1-57658

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TBC1 Domain Family Member 26
  • Antigen Symbol:
    TBC1D26
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    353149
  • Antigen Synonyms:
    MGC51025|member 26|TBC1 domain family member 26|TBC1 domain family
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to MGC51025 The peptide sequence was selected from the middle region of MGC51025. Peptide sequence RGRAWSLLLDIDRIKSQNPGKYKVMKEKGKRSSRIIHCIQLDVSHTLQKH.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102190-198
  • Supplier no.:
    NBP1-57658

Specifications

About this item

The TBC1D26 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBC1D26. This antibody reacts with human. The TBC1D26 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TBC1D26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human