Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ST6GALNAC1 Rabbit Polyclonal Antibody
Anti-ST6GALNAC1 Rabbit Polyclonal Antibody
Catalog # 102214-068
Supplier:  Novus Biologicals
Anti-ST6GALNAC1 Rabbit Polyclonal Antibody
Catalog # 102214-068
Supplier:  Novus Biologicals
Supplier Number:  NBP1-79210

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    ST6 (Alpha-N-Acetyl-Neuraminyl-2,3-Beta-Galactosyl-1,3)-N-Acetylgalactosaminide Alpha-2,6-Sialyltransferase 1
  • Antigen Symbol:
    ST6GALNAC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    55808
  • Antigen Synonyms:
    GalNAc alpha-2|ST6GalNAcIHSY11339|EC 2.4.99.3|SIAT7A|ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 1|6-sialyltransferase I|SIAT7-A|STYI|Sialyltransferase 7A|sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase) A|long form|alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1|GalNAc alpha-2,6-sialyltransferase I|ST6GalNAc I
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    The immunogen for this antibody is St6galnac1. Peptide sequence GALLLLTALHLCDKVSAYGFITEGHQRFSDHYYDTSWKRLIFYINHDFRL.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102214-068
  • Supplier no.:
    NBP1-79210

Specifications

About this item

The ST6GALNAC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST6GALNAC1. This antibody reacts with mouse. The ST6GALNAC1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ST6GALNAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse