To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sin3A-associated Protein, 30kDa
- Antigen Symbol:SAP30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:8819
- Antigen Synonyms:30 kDa Sin3-associated polypeptide|Sin3A-associated protein|Sin3-associated polypeptide p30|30kDa|histone deacetylase complex subunit SAP30|Sin3 corepressor complex subunit SAP30|sin3-associated polypeptide|30 kDa|Sin3-associated polypeptide
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen this antibody was made to, maps to a region between residue 170 and 220 of human Sin3A-Associated Protein, 30kDa using the numbering given in entry NP_003855.1 (GeneID 8819).
- Purification:Immunogen affinity purified
- Cat. No.:102217-812
- Supplier no.:NBP1-92648
Specifications
About this item
The SAP30 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SAP30. This antibody reacts with human, mouse. The SAP30 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: SAP30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse