Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Ras-related protein Rab-26
- Antigen Symbol:RAB26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:25837
- Antigen Synonyms:ras-related protein Rab-26|RAB26|V46133|member RAS oncogene family|Ras-related oncogene protein
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to RAB26(RAB26, member RAS oncogene family) The peptide sequence was selected from the N terminal of RAB26. Peptide sequence SRKKTPKSKGASTPAASTLPTANGARPARSGTALSGPDAPPNGPLQPGRP.
- Purification:Immunogen affinity purified
- Cat. No.:102191-488
- Supplier no.:NBP1-58882
The RAB26 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RAB26. This antibody reacts with human, mouse. The RAB26 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: RAB26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse