Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:pre-mRNA cleavage factor I (59 kDa subunit)
- Antigen Symbol:pre-mRNA cleavage Factor I (59 kDa Subunit)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:79869
- Antigen Synonyms:pre-mRNA cleavage factor I|CFIm59|59kDa|cleavage and polyadenylation specific factor 7|59 kDa subunit|FLJ12529,59 kDa subunit|CPSF 59 kDa subunit|FLJ39024|MGC9315
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cleavage and polyadenylation specificity factor 7, 59 kDa subunit using the numbering given in entry NP_079087.2 (GeneID 79869).
- Purification:Immunogen affinity purified
- Cat. No.:102111-256
- Supplier no.:NB100-61599
Specifications
About this item
The pre-mRNA cleavage factor I (59 kDa subunit) Antibody from Novus Biologicals is a rabbit polyclonal antibody to pre-mRNA cleavage factor I (59 kDa subunit). This antibody reacts with human. The pre-mRNA cleavage factor I (59 kDa subunit) Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: pre-mRNA cleavage factor I (59 kDa subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human