To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Phospholipase A2
- Antigen Symbol:PLA2G4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5321
- Antigen Synonyms:phosphatidylcholine 2-acylhydrolase|cPLA2|calcium-dependent phospholipid-binding protein|Phospholipase A2 group IVA|phospholipase A2|PLA2G4cytosolic phospholipase A2|calcium-dependent)|MGC126350|group IVA (cytosolic|cPLA2-alpha|lysophospholipase
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the N-terminus region of human PLA2G4A. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102164-500
- Supplier no.:NBP1-31344
The PLA2G4A Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLA2G4A. This antibody reacts with human, mouse. The PLA2G4A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:1000
Type: Primary
Antigen: PLA2G4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse