Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:p66 alpha
- Antigen Symbol:p66 alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.2 ml
- Gene ID:54815
- Antigen Synonyms:p66alpha|FLJ21017|p66 alpha|Hp66alpha|transcriptional repressor p66-alpha|GATA zinc finger domain containing 2A|GATA zinc finger domain-containing protein 2A|FLJ20085
- Storage Buffer:Whole antisera
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against a synthetic peptide corresponding to aa 137-150 of mouse p66a. It cross-reacts with human p66a (AAH19178.1).
- Purification:Unpurified
- Cat. No.:102110-198
- Supplier no.:NB100-56643
Specifications
About this item
The p66 alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to p66 alpha. This antibody reacts with human, mouse, rat. The p66 alpha Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: p66 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat