Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Nuclear Receptor Subfamily 2, Group E, Member 3
- Antigen Symbol:PNR/NR2E3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Hamster,Bat,Pig,Horse,Human,Bovine,Mouse
- Size:0.05 ml
- Gene ID:10002
- Antigen Synonyms:RNR|nuclear receptor subfamily 2|group E|member 3|Retina-specific nuclear receptor|PNRESCS|RP37MGC49976|photoreceptor-specific nuclear receptor|Nuclear receptor subfamily 2 group E member 3|rd7
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 19 amino acid peptide from ligand-binding domain of human NR2E3.
- Purification:Immunogen affinity purified
- Cat. No.:102253-414
- Supplier no.:NLS5770
Specifications
About this item
The PNR / NR2E3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PNR / NR2E3. This antibody reacts with human, mouse, porcine, bat, bovine, equine, hamster. The PNR / NR2E3 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PNR/NR2E3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Porcine, Bat, Bovine, Equine, Hamster