To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Nuclear Receptor Subfamily 2, Group C, Member 1
- Antigen Symbol:TR2/NR2C1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7181
- Antigen Synonyms:nuclear receptor subfamily 2 group C member 1|group C isoform|nuclear receptor subfamily 2|group C|Orphan nuclear receptor TR2|TR2-11|member 1|Testicular receptor 2|TR2|TR2 nuclear hormone receptor
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 1 and 50 of human Testicular Receptor 2 using the numbering given in entry NP_003288.2 (GeneID 7181).
- Purification:Immunogen affinity purified
- Cat. No.:102201-480
- Supplier no.:NBP1-71803
Specifications
About this item
The TR2 / NR2C1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TR2 / NR2C1. This antibody reacts with human, mouse. The TR2 / NR2C1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation.
Type: Primary
Antigen: TR2/NR2C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse