Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor Ionotropic N-methyl D-aspartate 2B
- Antigen Symbol:NMDAR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:2904
- Antigen Synonyms:NMDAR2BMGC142178|glutamate receptor|hNR3|N-methyl D-aspartate receptor subtype 2B|N-methyl-D-aspartate receptor subunit 3|ionotropic|NR3|MGC142180|N-methyl D-aspartate 2B|glutamate [NMDA] receptor subunit epsilon-2|NR2B|glutamate receptor subunit epsilon-2
- Modification:p Tyr1472
- Storage Buffer:10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phosphoTyr1472 of NMDA NR2B.
- Purification:Immunogen affinity purified
- Cat. No.:102130-128
- Supplier no.:NB300-182
Specifications
About this item
The NMDAR2B [p Tyr1472] Antibody from Novus Biologicals is a rabbit polyclonal antibody to NMDAR2B. This antibody reacts with human, mouse, rat. The NMDAR2B [p Tyr1472] Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat