Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor ionotropic N-methyl D-aspartate 1
- Antigen Symbol:NMDAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Gene ID:2902
- Antigen Synonyms:glutamate receptor|NMD-R1|ionotropic|NMDAR1glutamate [NMDA] receptor subunit zeta 1|subunit zeta-1|NR1|N-methyl D-aspartate 1|glutamate [NMDA] receptor subunit zeta-1|NMDA1|N-methyl-D-aspartate receptor subunit NR1|N-methyl-D-aspartate receptor channel
- Modification:p Ser896
- Storage Buffer:PBS [pH 7.4] containing 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+)
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human NMDAR1 around the phosphorylation site of serine 896 (R-R-SP-S-K).
- Purification:Immunogen affinity purified
- Cat. No.:102118-616
- Supplier no.:NB100-82092
The NMDAR1 [p Ser896] Antibody from Novus Biologicals is a rabbit polyclonal antibody to NMDAR1. This antibody reacts with human, mouse, rat. The NMDAR1 [p Ser896] Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: NMDAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat