Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Niemann-Pick type C1 Like-1
- Antigen Symbol:NPC1L1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:29881
- Antigen Synonyms:NPC1 (Niemann-Pick disease|NPC11L1|type C1|gene)-like 1|Niemann-Pick C1-like protein 1
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4 °C in the dark.
- Immunogen:A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 1000-1100). [UniProt# Q6T3U3]
- Purification:Immunogen affinity purified
- Cat. No.:102131-446
- Supplier no.:NB400-127G
Specifications
About this item
The Niemann-Pick type C1 Like-1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to Niemann-Pick type C1 Like-1. This antibody reacts with human, mouse, rat, porcine. The Niemann-Pick type C1 Like-1 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250
Type: Primary
Antigen: NPC1L1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine