Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa
- Antigen Symbol:NDUFV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:4723
- Antigen Synonyms:CI-51K|mitochondrial respiratory chain|Complex I-51kD|CI-51kD|UQOR1NADH dehydrogenase [ubiquinone] flavoprotein 1|EC 1.6.5.3|EC 1.6.99.5|FLJ59059|mitochondrial|51kDa|EC 1.6.99.3|NADH-ubiquinone oxidoreductase 51 kDa subunit|complex I|complex I 51kDa subunit|NADH dehydrogenase flavoprotein 1|mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1|CI51KD|NADH dehydrogenase (ubiquinone) flavoprotein 1 (51kD)|NADH dehydrogenase (ubiquinone) flavoprotein 1
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human NDUFV1. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102166-728
- Supplier no.:NBP1-33074
Rabbit Polyclonal NDUFV1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.
Type: Primary
Antigen: NDUFV1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human