To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa
- Antigen Symbol:NDUFC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4718
- Antigen Synonyms:B14.5b)|complex I subunit B14.5b|B14.5b|NADH dehydrogenase (ubiquinone) 1|NADH dehydrogenase [ubiquinone] 1 subunit C2|Human lung cancer oncogene 1 protein|2 (14.5kD|CI-B14.5b|NADHDH2|14.5kDa|Complex I-B14.5b|subcomplex unknown|HLC-1NADH dehydrogenase (ubiquinone) 1|2|NADH-ubiquinone oxidoreductase subunit B14.5b
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to NDUFC2(NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa) The peptide sequence was selected from the N terminal of NDUFC2. Peptide sequence IARRNPEPLRFLPDEARSLPPPKLTDPRLLYIGFLGYCSGLIDNLIR
- Purification:Immunogen affinity purified
- Cat. No.:102192-908
- Supplier no.:NBP1-59610
Rabbit Polyclonal NDUFC2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.
Type: Primary
Antigen: NDUFC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human