About this item
The pan Cadherin Antibody from Novus Biologicals is a rabbit polyclonal antibody to pan Cadherin. This antibody reacts with human, mouse, rat, amphibian, avian, bovine, canine, chicken, fish, rabbit, xenopus. The pan Cadherin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000
Type: Primary
Antigen: pan Cadherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Amphibian, Avian, Bovine, Canine, Chicken, Fish, Rabbit, Xenopus
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102129-552
- 102129-548
- Antigen Symbol:
- CDH2
- CDH2
- Antigen Name:
- pan Cadherin
- pan Cadherin
- Antigen Synonyms:
- cadherin-1|CD324|cadherin 1|ECAD|CD324 antigen|Epithelial cadherin|type 1|Arc-1|CAM 120/80|cell-CAM 120/80|UVOE-Cadherin|uvomorulin|E-cadherin (epithelial)|calcium-dependent adhesion protein|epithelial|LCAM|E-cadherin|CDHE
- cadherin-1|CD324|cadherin 1|ECAD|CD324 antigen|Epithelial cadherin|type 1|Arc-1|CAM 120/80|cell-CAM 120/80|UVOE-Cadherin|uvomorulin|E-cadherin (epithelial)|calcium-dependent adhesion protein|epithelial|LCAM|E-cadherin|CDHE
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Fish, Rabbit, Amphibian, Xenopus, Birds, Chicken, Human, Bovine, Dog, Rat, Mouse
- Fish, Rabbit, Amphibian, Xenopus, Birds, Chicken, Human, Bovine, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 999
- 999
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Synthetic peptide: DYDYLNDWGPRFKKLADMYGGGDD conjugated to KLH by a Glutaraldehyde linker. Sequence corrresponds to the C-terminal amino acids of N-Cadherin (Chicken), with an extra N-terminal lysine residue (24 amino acids).
- Synthetic peptide: DYDYLNDWGPRFKKLADMYGGGDD conjugated to KLH by a Glutaraldehyde linker. Sequence corrresponds to the C-terminal amino acids of N-Cadherin (Chicken), with an extra N-terminal lysine residue (24 amino acids).
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Whole antisera
- Whole antisera
- Storage Temperature:
- Store at -20C. Avoid freeze-thaw cycles.
- Store at -20C. Avoid freeze-thaw cycles.