Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MBD2 Rabbit Polyclonal Antibody
Anti-MBD2 Rabbit Polyclonal Antibody
Catalog # 102118-490
Supplier:  Novus Biologicals
Anti-MBD2 Rabbit Polyclonal Antibody
Catalog # 102118-490
Supplier:  Novus Biologicals
Supplier Number:  NB100-81657

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Methyl-CpG Binding Domain Protein 2
  • Antigen Symbol:
    MBD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    8932
  • Antigen Synonyms:
    methyl-CpG binding domain protein 2|NY-CO-41|demethylase|DKFZp586O0821|methyl-CpG-binding domain protein 2|Methyl-CpG-binding protein MBD2|DMTase
  • Storage Buffer:
    TBS and 0.1% BSA
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    0.2 mg/ml
  • Immunogen:
    The immunogen recognized by this antibody maps to a region between residue 361 to 411 of human methyl-CpG binding domain protein 2 using the numbering given in entry NP_003918.1 (GeneID 8932).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102118-490
  • Supplier no.:
    NB100-81657

Specifications

About this item

Rabbit Polyclonal MBD2 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MBD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human