Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:MAS1 oncogene
- Antigen Symbol:Mas
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4142
- Antigen Synonyms:MAS1 oncogene|proto-oncogene Mas|MGC119966|MAS
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to MAS1(MAS1 oncogene) The peptide sequence was selected from the middle region of MAS1. Peptide sequence VIIFIAILSFLVFTPLMLVSSTILVVKIRKNTWASHSSKLYIVIMVTIII.
- Purification:Immunogen affinity purified
- Cat. No.:102193-854
- Supplier no.:NBP1-60091
Rabbit Polyclonal MAS1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: MAS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human