To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 43 Member 1
- Antigen Symbol:LAT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:8501
- Antigen Synonyms:R00504|Solute carrier family 43 member 1|Prostate cancer overexpressed gene 1 protein|POV1large neutral amino acids transporter small subunit 3|PB39L-type amino acid transporter 3|member 1|LAT3prostate cancer overexpressed gene 1|solute carrier family 43
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to SLC43A1(solute carrier family 43, member 1) The peptide sequence was selected from the middle region of SLC43A1. Peptide sequence AVNKMLEYLVTGGQEHETNEQQQKVAETVGFYSSVFGAMQLLCLLTCPLI.
- Purification:Immunogen affinity purified
- Cat. No.:102192-554
- Supplier no.:NBP1-59431
Specifications
About this item
Rabbit Polyclonal LAT3 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.
Type: Primary
Antigen: SLC43A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human