Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Iron-responsive Element Binding Protein 2
- Antigen Symbol:IRP2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3658
- Antigen Synonyms:IRE-BP 2|ACO3|IRP2IRP2AD|EC 4.2.1.3|iron-responsive element-binding protein 2|iron-responsive element binding protein 2|Iron regulatory protein 2|FLJ23381
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to an internal portion of the murine IRP2 protein sequence (between residues 100-200). [UniProt Q811J3]
- Purification:Immunogen affinity purified
- Cat. No.:102106-006
- Supplier no.:NB100-1798H
Rabbit Polyclonal IRP2 Antibody [HRP]. Tested Applications: Western Blot. Tested Reactivity: Mouse.
Type: Primary
Antigen: IRP2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse