Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:3-hydroxy-3-methylglutaryl-CoA Synthase 1 (soluble)
- Antigen Symbol:HMGCS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:3157
- Antigen Synonyms:3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble)|HMGCS|cytoplasmic|HMG-CoA synthase|MGC90332|EC 2.3.3.10|hydroxymethylglutaryl-CoA synthase|3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble)|3-hydroxy-3-methylglutaryl coenzyme A synthase|3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to HMGCS1(3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble)) The peptide sequence was selected from the middle region of HMGCS1. Peptide sequence KDFTLNDFGFMIFHSPYCKLVQKSLARMLLNDFLNDQNRDKNSIYSGLEA.
- Purification:Immunogen affinity purified
- Cat. No.:102184-214
- Supplier no.:NBP1-54623
Specifications
About this item
Rabbit Polyclonal HMGCS1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: HMGCS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human