Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor 37 L1
- Antigen Symbol:GPR37L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:0.05 ml
- Gene ID:9283
- Antigen Synonyms:ET(B)R-LP-2|ETBR-LP-2endothelin type b receptor-like protein 2|G-protein coupled receptor 37-like 1|G-protein coupled receptor 37 like 1|endothelin B receptor-like protein 2|G protein-coupled receptor 37 like 1|ETBRLP2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR37L1.
- Purification:Immunogen affinity purified
- Cat. No.:102253-328
- Supplier no.:NLS405
Specifications
About this item
Rabbit Polyclonal GPR37L1 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Type: Primary
Antigen: GPR37L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human