Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:FLICE-associated huge protein
- Antigen Symbol:FLASH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9994
- Antigen Synonyms:FLASHKIAA1315CASP8-associated protein 2|RIP25FLASH homolog RIP25|CED-4|human FLASH|FLICE associated huge|FLICE-associated huge protein|caspase 8 associated protein 2|CASP8 associated protein 2|FLJ11208
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:C-terminus of the FLASH protein (proprietary peptide).
- Purification:Immunogen affinity purified
- Cat. No.:102132-016
- Supplier no.:NB500-219
Specifications
About this item
The FLASH Antibody from Novus Biologicals is a rabbit polyclonal antibody to FLASH. This antibody reacts with human. The FLASH Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: FLASH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human