Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DHX57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:90957
- Antigen Synonyms:DEAH box protein 57|DEAH-box RNA/DNA helicase AAM73547|FLJ32861|EC 3.6.4.13|DDX57|EC 3.6.1|DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57|putative ATP-dependent RNA helicase DHX57
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 150 and200 of human DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 using the numbering given in entry NP_945314.1 (GeneID 90957).
- Purification:Immunogen affinity purified
- Cat. No.:102162-618
- Supplier no.:NBP1-28738
Specifications
About this item
The DHX57 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DHX57. This antibody reacts with human. The DHX57 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: DHX57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human