To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Calcium Channel, Voltage-dependent, Alpha 2/delta Subunit 4
- Antigen Symbol:CACNA2D4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:93589
- Antigen Synonyms:RCD4|Voltage-gated calcium channel subunit alpha-2/delta-4|calcium channel|voltage-gated calcium channel alpha(2)delta-4 subunit|alpha 2/delta subunit 4|voltage-dependent|voltage-dependent calcium channel subunit alpha-2/delta-4
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:The immunogen for this antibody is Cacna2d4 - middle region. Peptide sequence DYVHYIEPCFKGILVQADRDNREHFKQLVDELMVKGVGVVSQALIEAFEI.
- Purification:Immunogen affinity purified
- Cat. No.:102223-254
- Supplier no.:NBP1-98469
The CACNA2D4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CACNA2D4. This antibody reacts with mouse. The CACNA2D4 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CACNA2D4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse