Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CSAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:126731
- Antigen Synonyms:chromosome 1 open reading frame 96|cilia and spindle-associated protein|RP4-803J11.3|C1orf96|centriole
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to C1orf96 (chromosome 1 open reading frame 96) The peptide sequence was selected from the middle region of C1orf96. Peptide sequence ENKHPFALYGWGEKQTDTGSQKTHNVCASAPVHEIHESALRAKNRRQVEK.
- Purification:Immunogen affinity purified
- Cat. No.:102199-686
- Supplier no.:NBP1-70458
The CSAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to CSAP. This antibody reacts with human. The CSAP Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CSAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human