Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:SWI/SNF related matrix associated actin dependent Regulator of chromatin subfamily a member 4
- Antigen Symbol:Brg1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:6597
- Antigen Synonyms:BRM/SWI2-related gene 1|SNF2B|brahma protein-like 1|SWI/SNF related|Protein brahma homolog 1|BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4|sucrose nonfermenting-like 4|ATP-dependent helicase SMARCA4|SWI2|homeotic gene regulator|SNF2-BETA|transcription activator BRG1|FLJ39786|SNF2L4|member 4|SNF2|global transcription activator homologous sequence|hSNF2b|Protein BRG-1|BAF190|nuclear protein GRB1|actin dependent regulator of chromatin|RTPS2|EC 3.6.1|subfamily a|SNF2-like 4|SNF2LB|EC 3.6.4.-|BAF190A|matrix associated|Mitotic growth and transcription activator|BRG1-associated factor 190A
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human BRG1/SMARCA4 using the numbering given in entry NP_003063.2 (GeneID 6597).
- Purification:Immunogen affinity purified
- Cat. No.:102107-854
- Supplier no.:NB100-2594
The Brg1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Brg1. This antibody reacts with human, mouse. The Brg1 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.
Type: Primary
Antigen: Brg1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse