Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 2
- Antigen Symbol:AOC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:314
- Antigen Synonyms:RAOEC 1.4.3.21|EC 1.4.3|Semicarbazide-sensitive amine oxidase|Amine oxidase [copper-containing]|copper containing 2 (retina-specific)|retina-specific copper amine oxidase|SSAO|DAO2|amine oxidase
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to AOC2(amine oxidase, copper containing 2 (retina-specific)) The peptide sequence was selected from the middle region of AOC2 (NP_033720). Peptide sequence QATMVDIHILVGKGAVQLLPGAVCVFEEAQGLPLRRHHNYLQNHFYGGLA.
- Purification:Immunogen affinity purified
- Cat. No.:102195-606
- Supplier no.:NBP1-62442
Specifications
About this item
The AOC2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AOC2. This antibody reacts with human. The AOC2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: AOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human